Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR5/FGFRL1 Antibody (004), PE, Novus Biologicals™
SDP

Catalog No. NB333148 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333148 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333148 Supplier Novus Biologicals Supplier No. NBP290466PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

FGFR5/FGFRL1 Monoclonal antibody specifically detects FGFR5/FGFRL1 in Mouse samples. It is validated for ELISA

Specifications

Antigen FGFR5/FGFRL1
Applications ELISA
Classification Monoclonal
Clone 004
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias FGF homologous factor receptor, FGF receptor-like protein 1, FGFR5, FGFR-5, FGFR-like protein, FHFR, Fibroblast growth factor receptor 5, fibroblast growth factor receptor-like 1
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse FGFR5/FGFRL1 (Accession#: NP_473412.1, Met 1-Pro 374)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 53834
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.