Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR5/FGFRL1 Antibody (RM0232-7L43), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP212294IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212294IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212294IR Supplier Novus Biologicals Supplier No. NBP212294IR
Add to Cart
Add to Cart

Rat Monoclonal Antibody

FGFR5/FGFRL1 Monoclonal specifically detects FGFR5/FGFRL1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen FGFR5/FGFRL1
Applications Western Blot
Classification Monoclonal
Clone RM0232-7L43
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FGF homologous factor receptor, FGF receptor-like protein 1, FGFR5, FGFR-5, FGFR-like protein, FHFR, Fibroblast growth factor receptor 5, fibroblast growth factor receptor-like 1
Gene Symbols FGFRL1
Host Species Rat
Immunogen Mouse recombinant FGFR5/FGFRL1
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 53834
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.