Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGGY carbohydrate kinase domain containing Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093993 100 μL
NB093994 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093993 Supplier Novus Biologicals Supplier No. NBP303388100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FGGY carbohydrate kinase domain containing Polyclonal specifically detects FGGY carbohydrate kinase domain containing in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen FGGY carbohydrate kinase domain containing
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias EC 2.7.1, EC 2.7.1.-, FGGY carbohydrate kinase domain containing, FGGY carbohydrate kinase domain-containing protein, FLJ10986
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human FGGY carbohydrate kinase domain containing (NP_060761.3).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 55277
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.