Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGL2/Fibroleukin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | FGL2/Fibroleukin |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15696520
![]() |
Novus Biologicals
NBP15696520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP156965
![]() |
Novus Biologicals
NBP156965 |
100 μL |
Each for $487.50
|
|
|||||
Description
FGL2/Fibroleukin Polyclonal specifically detects FGL2/Fibroleukin in Human samples. It is validated for Western Blot.Specifications
FGL2/Fibroleukin | |
Polyclonal | |
Rabbit | |
Cancer | |
fibrinogen-like 2, Fibrinogen-like protein 2, fibroleukin, pT49T49 | |
FGL2 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Q14314 | |
10875 | |
Synthetic peptides corresponding to FGL2 (fibrinogen-like 2) The peptide sequence was selected from the middle region of FGL2. Peptide sequence WTVLQARLDGSTNFTRTWQDYKAGFGNLRREFWLGNDKIHLLTKSKEMIL. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title