Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

fHbp Mouse anti-Bacteria, Clone: JAR 5, MilliporeSigma™

Mouse Monoclonal Antibody

$191.34 - $481.79

Specifications

Antigen fHbp
Clone JAR 5
Concentration 0.5 mg/mL
Applications Dot Blot, ELISA, Inhibition Assays, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF266925
View Documents
MilliporeSigma
MABF266925UL
25 μL
Each for $191.34
Only null left
Add to Cart
 
MABF2669100
View Documents
MilliporeSigma
MABF2669100UL
100 μL
Each for $481.79
Only null left
Add to Cart
 
Description

Description

fHbp Monoclonal antibody specifically detects fHbp in Bacteria samples. It is validated for Dot Blot, ELISA, Inhibition, Western Blotting.
Specifications

Specifications

fHbp
0.5 mg/mL
Monoclonal
Purified
RUO
Purified mouse monoclonal antibody IgG2b in PBS without azide.
IgG2b κ
Protein G purified
JAR 5
Dot Blot, ELISA, Inhibition Assays, Western Blot
Unconjugated
Mouse
Bacteria
His-tagged recombinant Factor H-binding protein (fHbp) from variant group 1, strain MC58, expressed in E. coli.
Primary
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.