Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FHIT Antibody (104), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB100436 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100436 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100436 Supplier Novus Biologicals Supplier No. NBP290283IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

FHIT Monoclonal specifically detects FHIT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FHIT
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 104
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias AP3A hydrolase, AP3AaseFRA3Bbis(5'-adenosyl)-triphosphatase, Diadenosine 5'-5'''-P1, dinucleosidetriphosphatase, EC 3.6.1.29, fragile histidine triad gene, Fragile histidine triad protein, P3-triphosphate hydrolase, tumor suppressor protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human FHIT (P49789; Met1-Gln147).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Lipid and Metabolism, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2272
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.