Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FHIT Antibody (104), FITC, Novus Biologicals™
SDP

Catalog No. NB038530 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB038530 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB038530 Supplier Novus Biologicals Supplier No. NBP290283F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

FHIT Monoclonal specifically detects FHIT in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FHIT
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 104
Conjugate FITC
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias AP3A hydrolase, AP3AaseFRA3Bbis(5'-adenosyl)-triphosphatase, Diadenosine 5'-5'''-P1, dinucleosidetriphosphatase, EC 3.6.1.29, fragile histidine triad gene, Fragile histidine triad protein, P3-triphosphate hydrolase, tumor suppressor protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human FHIT (P49789; Met1-Gln147).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Lipid and Metabolism, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2272
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.