Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FHL1 Antibody (OTI3G7), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB270739A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270739A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270739A48 Supplier Novus Biologicals Supplier No. NBP270739AF488

Mouse Monoclonal Antibody

FHL1 Monoclonal specifically detects FHL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FHL1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3G7
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bA535K18.1, four and a half LIM domains 1, KYOT, KYO-T, LIM protein SLIMMER, skeletal muscle LIM-protein1, SLIM1Isl-1 and Mec-3 domains 1
Gene Symbols FHL1
Host Species Mouse
Immunogen Full length recombinant protein of human FHL1 (NP_001440) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Lipid and Metabolism, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2273.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.