Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FHL2 Antibody (AT21D11), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB163283 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163283 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163283 Supplier Novus Biologicals Supplier No. NBP261162AF350
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FHL2 Monoclonal antibody specifically detects FHL2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen FHL2
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone AT21D11
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias down-regulated in rhabdomyosarcoma LIM protein, DRALAAG11, FHL-2, four and a half LIM domains 2, four and a half LIM domains protein 2, four and a half LIM-domain protein 2, LIM domain protein DRAL, Skeletal muscle LIM-protein 3, SLIM-3, SLIM3aging-associated gene 11
Host Species Mouse
Immunogen Recombinant human FHL2 (1-279aa) purified from E. coli.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2274
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.