Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FHL5 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB360591 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360591 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360591 Supplier Novus Biologicals Supplier No. NBP227588APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FHL5 Polyclonal antibody specifically detects FHL5 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen FHL5
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias activator of cAMP-responsive element modulator (CREM) in testis, Activator of cAMP-responsive element modulator in testis, Activator of CREM in testis, ACTKIAA0776, dJ393D12.2, FHL-5, FLJ33049, four and a half LIM domains 5,1700027G07Rik, four and a half LIM domains protein 5, LIM protein ACT, RP3-393D12.2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminus of ACT protein
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9457
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.