Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FHL5 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP227588 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227588 0.1 mg
NB019415 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227588 Supplier Novus Biologicals Supplier No. NBP227588
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FHL5 Polyclonal specifically detects FHL5 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen FHL5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q5TD97, Q9WTX7, Q6AXT1
Gene Alias activator of cAMP-responsive element modulator (CREM) in testis, Activator of cAMP-responsive element modulator in testis, Activator of CREM in testis, ACTKIAA0776, dJ393D12.2, FHL-5, FLJ33049, four and a half LIM domains 5,1700027G07Rik, four and a half LIM domains protein 5, LIM protein ACT, RP3-393D12.2
Gene Symbols FHL5
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminus of ACT protein
Molecular Weight of Antigen 32.7 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9457
Test Specificity Human and Mouse. Based upon immunogen sequence homology, this antibody is predicted to react with Rat (97%) and Bovine (90%).
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.