Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FHOD3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224544 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224544 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224544 Supplier Novus Biologicals Supplier No. NBP224544
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FHOD3 Polyclonal specifically detects FHOD3 in Human, Mouse, Bovine, Canine, Chicken, Equine, Opossum samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FHOD3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 15 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q2V2M9
Gene Alias FH1/FH2 domain-containing protein 3, FHOS2, formactin-2, Formactin2, formin homolog overexpressed in spleen 2, formin homology 2 domain containing 3, hFHOS2, KIAA1695
Gene Symbols FHOD3
Host Species Rabbit
Immunogen A portion of amino acids 1250-1300 of human FHOD3 (CRA-d) was used as the immunogen.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80206
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform 1 and isoform 2), rat, mouse, zebrafish, xenopus and bovine.
Target Species Human, Mouse, Bovine, Canine, Chicken, Equine, Opossum
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.