Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrillarin Antibody (28F2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105223
Description
Fibrillarin Monoclonal specifically detects Fibrillarin in Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| Fibrillarin | |
| Monoclonal | |
| Unconjugated | |
| Supplied as concentrated hybridoma cell culture media. with 5mM Sodium Azide | |
| FBL | |
| Yeast nuclear preparation (S. cerevisiae). | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| Yeast | |
| Supernatant |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 28F2 | |
| Western Blot 1:10000 (cell lysates)-1:25000 (nuclear fractions), Immunocytochemistry/Immunofluorescence 1:2000, Immunoprecipitation | |
| EC 2.1.1,34-kD nucleolar scleroderma antigen, EC 2.1.1.-, EC 2.1.1.37, FIB, FIB1,34 kDa nucleolar scleroderma antigen, fibrillarin, FLRNrRNA 2'-O-methyltransferase fibrillarin, RNA, U3 small nucleolar interacting protein 1, RNU3IP1 | |
| Mouse | |
| 34.5 kDa | |
| 0.5 mL | |
| Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
| 2091 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction