Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrillarin Antibody (28F2), Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
| Antigen | Fibrillarin |
|---|---|
| Clone | 28F2 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Supernatant |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP105223
![]() |
Novus Biologicals
NBP105223 |
0.5 mL | N/A | N/A | N/A | ||||
Description
Fibrillarin Monoclonal specifically detects Fibrillarin in Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| Fibrillarin | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Yeast | |
| 2091 | |
| Yeast nuclear preparation (S. cerevisiae). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 28F2 | |
| Unconjugated | |
| Mouse | |
| Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
| EC 2.1.1,34-kD nucleolar scleroderma antigen, EC 2.1.1.-, EC 2.1.1.37, FIB, FIB1,34 kDa nucleolar scleroderma antigen, fibrillarin, FLRNrRNA 2'-O-methyltransferase fibrillarin, RNA, U3 small nucleolar interacting protein 1, RNU3IP1 | |
| FBL | |
| IgG3 | |
| Tissue culture supernatant | |
| 34.5 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title