Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrillarin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25859225UL
Description
Fibrillarin Polyclonal specifically detects Fibrillarin in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Fibrillarin | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
EC 2.1.1,34-kD nucleolar scleroderma antigen, EC 2.1.1.-, EC 2.1.1.37, FIB, FIB1,34 kDa nucleolar scleroderma antigen, fibrillarin, FLRNrRNA 2'-O-methyltransferase fibrillarin, RNA, U3 small nucleolar interacting protein 1, RNU3IP1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
FBL | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EYRAWNPFRSKLAAAILGGVDQIHIKP | |
25 μL | |
Cellular Markers, Core ESC Like Genes, Neuroscience, Nuclear Envelope Markers, Stem Cell Markers | |
2091 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction