Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrillarin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Fibrillarin |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Fibrillarin Polyclonal specifically detects Fibrillarin in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Fibrillarin | |
Polyclonal | |
Rabbit | |
Cellular Markers, Core ESC Like Genes, Neuroscience, Nuclear Envelope Markers, Stem Cell Markers | |
EC 2.1.1,34-kD nucleolar scleroderma antigen, EC 2.1.1.-, EC 2.1.1.37, FIB, FIB1,34 kDa nucleolar scleroderma antigen, fibrillarin, FLRNrRNA 2'-O-methyltransferase fibrillarin, RNA, U3 small nucleolar interacting protein 1, RNU3IP1 | |
FBL | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
2091 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EYRAWNPFRSKLAAAILGGVDQIHIKP | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title