Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrillarin Antibody, Novus Biologicals™
SDP

Catalog No. NB019693 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019693 0.025 mL
NBP246881 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019693 Supplier Novus Biologicals Supplier No. NBP2468810.025ML
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Fibrillarin Polyclonal specifically detects Fibrillarin in Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Fibrillarin
Applications Immunofluorescence, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:5000, Immunocytochemistry/Immunofluorescence 1:2000-1:5000
Formulation PBS with 5mM Sodium Azide
Gene Accession No. P22087
Gene Alias EC 2.1.1,34-kD nucleolar scleroderma antigen, EC 2.1.1.-, EC 2.1.1.37, FIB, FIB1,34 kDa nucleolar scleroderma antigen, fibrillarin, FLRNrRNA 2'-O-methyltransferase fibrillarin, RNA, U3 small nucleolar interacting protein 1, RNU3IP1
Gene Symbols FBL
Host Species Chicken
Immunogen Full length human Fibrillarin expressed in and purified from E.Coli. [UniProt# P22087]
Molecular Weight of Antigen 34.5 kDa
Purification Method IgY purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Core ESC Like Genes, Neuroscience, Nuclear Envelope Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2091
Target Species Human, Mouse, Bovine, Chicken, Equine, Rat, Pig
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.