Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrin Antibody (UC45), Novus Biologicals™
SDP

Catalog No. NBP250407 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250407 0.1 mL
NB395434 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP250407 Supplier Novus Biologicals Supplier No. NBP250407
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fibrin Monoclonal antibody specifically detects Fibrin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmune Assays, CyTOF

Specifications

Antigen Fibrin
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA), CyTOF
Classification Monoclonal
Clone UC45
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000, CyTOF-ready
Formulation PBS
Host Species Mouse
Immunogen Human acute monoblastic leukemia cells.
Purification Method Thiophilic Chromatography
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.