Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrin beta-chain Antibody (8E5), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB342801 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342801 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342801 Supplier Novus Biologicals Supplier No. NBP281110MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fibrin beta-chain Monoclonal antibody specifically detects Fibrin beta-chain in Human samples. It is validated for ELISA

Specifications

Antigen Fibrin beta-chain
Applications ELISA
Classification Monoclonal
Clone 8E5
Conjugate mFluor Violet 450 SE
Dilution ELISA, In vivo assay
Formulation 50mM Sodium Borate
Gene Alias Beta-Fibrinogen, epididymis secretory sperm binding protein Li 78p, fibrinogen beta chain, fibrinogen, B beta polypeptide, HEL-S-78p
Host Species Mouse
Immunogen 8E5 was prepared fusing Sp2/o myeloma cells with spleen cells derived from female BALB/c mice immunized with a synthetic beta-fibrin peptide seqeunce GHRPLDKC conjugated to MB-KLH (maleimidobenzoylated keyhole limpet hemocyanin).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 2244
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.