Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrin Mouse anti-Human,Mouse, KHL, Clone: 59D8, MilliporeSigma™
Mouse Monoclonal Antibody
$190.80 - $499.00
Specifications
Antigen | Fibrin |
---|---|
Clone | 59D8 |
Applications | Immunofluorescence, Immunohistochemistry, Inhibition Assays, Radioimmune Assays (RIA), Western Blot |
Classification | Monoclonal |
Conjugate | KLH |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS21552MI
|
MilliporeSigma
MABS215525UG |
25 μg |
Each for $190.80
|
|
|||||
MABS21551MI
|
MilliporeSigma
MABS2155100UG |
100 μg |
Each for $499.00
|
|
|||||
Description
Fibrin Monoclonal antibody specifically detects Fibrin in Human,Mouse samples. It is validated for Immunofluorescence, Immunohistochemistry, Peptide Inhibition Assay, Radioimmunoassay, Western Blotting.Specifications
Fibrin | |
Immunofluorescence, Immunohistochemistry, Inhibition Assays, Radioimmune Assays (RIA), Western Blot | |
KLH | |
Mouse | |
Human, Mouse | |
Fibrinogen beta chain | |
KLH-conjugated synthetic heptapeptide (GHRPLNK) derived from the N-terminal region of the beta chain of human fibrinogen. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼55 kDa observed; 55.93 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
59D8 | |
Monoclonal | |
Purified | |
Signaling | |
NP_001171670 | |
FGB | |
IgG1 κ | |
Protein G Purified | |
Clone 59D8 is a mouse monoclonal antibody that specifically detects human and murine fibrin. It targets an epitope within a heptapeptide fom the N-terminal region of the beta chain. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title