Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrinogen gamma chain Antibody (OTI2D2), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP270732V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270732V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270732V Supplier Novus Biologicals Supplier No. NBP270732V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fibrinogen gamma chain Monoclonal specifically detects Fibrinogen gamma chain in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Fibrinogen gamma chain
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2D2
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias fibrinogen gamma chain, fibrinogen, gamma polypeptide
Gene Symbols FGG
Host Species Mouse
Immunogen Full length recombinant protein of human Fibrinogen gamma chain (NP_000500) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2266.0
Target Species Human, Mouse, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.