Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrinogen Mouse anti-Human, Clone: UC45, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30722920UG
Description
Fibrinogen Monoclonal specifically detects Fibrinogen in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Fibrinogen | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml | |
Fib2, fibrinogen alpha chain, fibrinogen, A alpha polypeptide, MGC119422, MGC119423, MGC119425 | |
Human acute monoblastic leukemia cells. | |
20 μg | |
Cell Cycle and Replication | |
2243 | |
Store at 4°C. | |
IgM κ |
Flow Cytometry, ELISA, Immunofluorescence | |
UC45 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein L purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction