Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrinogen Mouse anti-Human, Clone: UC45, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$280.00

Specifications

Antigen Fibrinogen
Clone UC45
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications Flow Cytometry, ELISA, Immunofluorescence
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121932
SDP
View Documents
Novus Biologicals
NBP30722920UG
20 μg
Each of 1 for $280.00
Only null left
Add to Cart
 
Description

Description

Fibrinogen Monoclonal specifically detects Fibrinogen in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Fibrinogen
0.2 mg/ml
Flow Cytometry, ELISA, Immunofluorescence
Unconjugated
Mouse
Cell Cycle and Replication
10 mM PBS with 0.05% BSA
2243
IgM κ
Protein L purified
UC45
Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Monoclonal
Purified
RUO
Human
Fib2, fibrinogen alpha chain, fibrinogen, A alpha polypeptide, MGC119422, MGC119423, MGC119425
Human acute monoblastic leukemia cells.
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.