Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrinopeptide B, Mouse anti-Human, Clone: 800000, Novus Biologicals™
SDP

Catalog No. NB016304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016304 0.025 mg
NB016305 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB016304 Supplier Novus Biologicals Supplier No. NBP2811100.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fibrin beta-chain Monoclonal specifically detects Fibrin beta-chain in Human samples. It is validated for ELISA, In vivo assay.

Specifications

Antigen Fibrinopeptide B
Applications ELISA
Classification Monoclonal
Clone 800000
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, In vivo assay
Formulation PBS
Gene Alias fibrinogen beta chain, fibrinogen, B beta polypeptide, MGC104327, MGC120405
Gene Symbols FGB
Host Species Mouse
Immunogen 8E5 was prepared fusing Sp2/o myeloma cells with spleen cells derived from female BALB/c mice immunized with a synthetic beta-fibrin peptide seqeunce GHRPLDKC conjugated to MB-KLH (maleimidobenzoylated keyhole limpet hemocyanin).
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 2244
Test Specificity 8E5 binds to the amino-terminal region of the beta-chain of human fibrin at an epitope consiting of GHRPLDKC. Binding is possible in both the monomeric form and the polymeric form of the protein. This peptide sequence is found only in fibrin, and not in its precursor fibrinogen and so there is no cross-reactivity with fibrinogen. Fibrin is a non-globular protein which plays a role in the clotting of blood and circulates as the precursor fibrinogen. Polymerization occurs through the action of the protease thrombin, which results in the active fibrin. Fibrin polymerizes as an insoluble matrix and forms a clot along with platelets over a wound site.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.