Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibroblast Activation Protein alpha/FAP Antibody (427819), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB312800 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312800 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312800 Supplier Novus Biologicals Supplier No. FAB3715APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fibroblast Activation Protein alpha/FAP Monoclonal antibody specifically detects Fibroblast Activation Protein alpha/FAP in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Fibroblast Activation Protein alpha/FAP
Applications Flow Cytometry
Classification Monoclonal
Clone 427819
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Fibroblast Activation Protein alpha/FAP, Leu26-Asp760, Accession # Q12884
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 2191
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.