Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibroblast Activation Protein alpha/FAP Antibody (983802), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB302340 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302340 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302340 Supplier Novus Biologicals Supplier No. FAB9727MFV450
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Fibroblast Activation Protein alpha/FAP Monoclonal specifically detects Fibroblast Activation Protein alpha/FAP in Mouse samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Fibroblast Activation Protein alpha/FAP
Applications ELISA, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 983802
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, ELISA, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Fibroblast Activation Protein alpha/FAP, Leu26-Asp761, Accession # P97321
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 2191
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.