Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibroblast Antibody (ER-TR7) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013809 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013809 0.125 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013809 Supplier Novus Biologicals Supplier No. NBP280736
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Fibroblast Monoclonal specifically detects Fibroblast in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, CyTOF-ready.

Specifications

Antigen Fibroblast
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), CyTOF
Classification Monoclonal
Clone ER-TR7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 1:10-1:1000, Immunohistochemistry reported in scientific literature (PMID 22042977), Immunocytochemistry/Immunofluorescence : 1:10-1:500, Immunohistochemistry-Frozen : 1:500, CyTOF-ready
Formulation PBS
Gene Alias reticular fibroblast
Host Species Rat
Immunogen Isolated C3H thymic stromal cells
Purification Method Protein G purified
Quantity 0.125 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.