Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibroblast Antibody (ER-TR7), PE, Novus Biologicals™
SDP

Catalog No. NB10064932Q Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064932Q 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064932Q Supplier Novus Biologicals Supplier No. NB10064932PE
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Fibroblast Monoclonal specifically detects Fibroblast in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).

Specifications

Antigen Fibroblast
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ER-TR7
Conjugate PE
Dilution Flow Cytometry
Gene Alias reticular fibroblast
Host Species Rat
Immunogen Isolated C3H thymic stromal cells
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64932 recognizes ER-TR7, an antigen that is located in the cytoplasm of reticular fibroblasts and is a component of the extracellular matrix of lymphoid and non-lymphoid organs. The antigen recognized by clone ER-TR7 has not been identified but studies suggest that it is likely to be distinct from laminin, fibronectin, collagen types I-IV, heparin sulphate proteoglycan, entactin and nidogen. Clone ER-TR7 has been used to stain the microanatomy of various organs and also stains subendothelial deposits in atherosclerotic plaques.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.