Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibroblast Surface Protein Antibody (1B10), Novus Biologicals™
SDP

Catalog No. NB1001845 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001845 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001845 Supplier Novus Biologicals Supplier No. NB1001845
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Fibroblast Surface Protein Monoclonal specifically detects Fibroblast Surface Protein in Human, Mouse samples. It is validated for Complement-dependent Cytotoxicity, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen Fibroblast Surface Protein
Applications Complement-dependent Cytotoxicity, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 1B10
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10-1:500, Complement-dependent Cytotoxicity 1:500
Formulation The product is provided as the immunoglobulin fraction of mouse ascites fluid in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA. with 15mM Sodium Azide
Host Species Mouse
Immunogen cultured human thymic fibroblasts
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Fibroblast Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Test Specificity This fibroblast antigen has been demonstrated on human synovial, mammary, foreskin and thymic fibroblasts and in malignant fibrosarcoma tissue. The antigen is both cell membrane and lysosome associated.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.