Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibroblasts/Epithelial cells Antibody (D7-FIB), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB60777A594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB60777A594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB60777A594 Supplier Novus Biologicals Supplier No. NB600777AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fibroblasts/Epithelial cells Monoclonal specifically detects Fibroblasts/Epithelial cells in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen Fibroblasts/Epithelial cells
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone D7-FIB
Conjugate Alexa Fluor 594
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen Human foreskin fibroblasts
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity NB600-777 recognizes a 112kD molecule expressed on the cell surface of human fibroblasts and epithelial cells. In peripheral blood the antibody stains myeloid cells and a very small number of lymphocytes. Studies upon the antigen have shown it to be sensitive to SDS, but resistant to trypsin, tunicamycin and collagenase. In immunohistological studies the antibody has also been found to bind to epithelium, myoepthelium, smooth muscle and some leucocytes. D7-FIB has been shown to be useful as a cell membrane marker to characterize chondrocyte differentiation giving a positive reaction with dedifferentiated human chondrocytes, and negative with differentiated chondrocytes (3). This product is routinely tested in flow cytometry on the KG1 cell line.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.