Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin Antibody (014), PE, Novus Biologicals™
SDP

Catalog No. NB307684 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307684 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307684 Supplier Novus Biologicals Supplier No. NBP289407PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Fibronectin Monoclonal antibody specifically detects Fibronectin in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen Fibronectin
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 014
Conjugate PE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation PBS
Gene Alias CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Fibronectin (Ser 607-Pro 1265; Accession#: NP_997647.1).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2335
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.