Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin Antibody (960632), DyLight 350, R&D Systems™
SDP

Catalog No. MAB8258UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8258UV 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8258UV Supplier Novus Biologicals Supplier No. MAB8258UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Immunohistochemistry, Western Blot.

Specifications

Antigen Fibronectin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 960632
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
Gene Symbols FN1
Host Species Mouse
Immunogen Partial recombinant human Fibronectin/Anastellin protein (amino acids 631-705) [UniProt P02751]
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2335
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.