Learn More
Description
Specifications
Specifications
| Antigen | Fibronectin |
| Applications | Western Blot, ELISA, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | A2 |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Gene Alias | CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF |
| Host Species | Rabbit |
| Immunogen | 2D-PAGE gel isolated human Extra-Domain containing Fibronectin from lysate of human melanoma COLO-38 cells. |
| Purification Method | Protein A purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
