Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibronectin Antibody (A32) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2235250.1ML
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Fibronectin | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
FN1 | |
Bovine corneal endothelial cells | |
0.1 mL | |
Primary | |
Clone A32 is highly specific for fibronectin. There is no evidence for cross-reactivity with other connective tissue proteins (vitronectin, elastin, collagen, laminin). Clone A32 cross-reacts with human fibronectin. Other species have not been tested. | |
Store at 4C. Do not freeze. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
A32 | |
Unconjugated | |
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
Mouse | |
Protein A or G purified | |
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
2335 | |
Human, Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction