Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin Antibody (A32) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$536.00

Specifications

Antigen Fibronectin
Clone A32
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP22352501
SDP
View Documents
Novus Biologicals
NBP2235250.1ML
0.1 mL
Each of 1 for $536.00
Only null left
Add to Cart
 
Description

Description

Fibronectin Monoclonal specifically detects Fibronectin in Human, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Fibronectin
1 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
Mouse
Human, Bovine
2335
Bovine corneal endothelial cells
Primary
Store at 4C. Do not freeze.
A32
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Monoclonal
Purified
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
FN1
IgG1 κ
Protein A or G purified
Clone A32 is highly specific for fibronectin. There is no evidence for cross-reactivity with other connective tissue proteins (vitronectin, elastin, collagen, laminin). Clone A32 cross-reacts with human fibronectin. Other species have not been tested.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.