Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Fibronectin Antibody Blocking Peptide
Shop All Novus Biologicals Products

Click to view available options
Quantity:
0.1 mg
Specifications
Specifications
Gene ID (Entrez) | 2335 |
Concentration | 1 mg/ml |
Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
For Use With (Application) | This peptide is useful as a blocking peptide for NBP1-91258. For further blocking peptide related protocol, click here . |
Gene Alias | CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF |
Gene Symbol | FN1 |
Quantity | 0.1 mg |
Research Category | Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers |
Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction