Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin Antibody (DH1), Novus Biologicals™
SDP

Catalog No. NBP151723 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP151723 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP151723 Supplier Novus Biologicals Supplier No. NBP151723
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fibronectin Monoclonal specifically detects Fibronectin in Human, Rat, Chicken, Guinea Pig, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Fibronectin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DH1
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Gene Alias CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
Gene Symbols FN1
Host Species Mouse
Immunogen The antibody reacts with Fibronectin.
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2335
Test Specificity The antibody is specific to extradomain A (EDA) sequence of a cellular fibronectin and recognizes thus only the cellular fibronectin. Mab DHI has been shown to specifically block chondrocyte condensation in chicken embryos (5). The antibody cross reacts with human, chicken, rat, rabbit and guinea pig. Fibronectins are diverse high molecular weight cell adhesion glycoproteins found in basement membranes, in the interstitial connective tissue matrix, and in soluble in plasma and other body fluids. In adult tissues most fibronectin is produced by hepatocytes (pFn) that differ from locally produced cellular fibronectins. A monoclonal antibody (BF12) has been produced to fibronectin that reacts with both the cellular and plasma forms of the protein and recognizes the cell-binding region in fibronectin. The monoclonal antibody DH1 recognizes only cellular form of Fibronectin. DH1 is specific to the extra domain A (EDA) of the molecule. cFns are found in some basement membranes during development and only in distinct tissues in adult, for instance in smooth muscle and alveolar walls of airways. cFns are abundant in the plasma of carcinoma patients and in the stroma of various carcinomas. Monoclonal antibody (Mab) to cFn is derived from the hybridoma produced by fusion between myeloma cells and Balb/c spleen cells. Purified fibronectin from A8387 fibrosarcoma cells was used as an immunogen (21).
Target Species Human, Rat, Chicken, Guinea Pig, Rabbit
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.