Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin Antibody (Fn-3), Janelia Fluor™ 646
SDP

Catalog No. NB002652646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002652646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002652646 Supplier Novus Biologicals Supplier No. NB1002652JF646

Mouse Monoclonal Antibody

Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Fibronectin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone Fn-3
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
Gene Symbols FN1
Host Species Mouse
Immunogen FR5 cells, derived by SV40 transformation of human mammary epithelial cells.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2335
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.