Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin Antibody (HFN7.1), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP234633W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234633W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234633W Supplier Novus Biologicals Supplier No. NBP234633AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fibronectin Monoclonal specifically detects Fibronectin in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Fibronectin
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HFN7.1
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
Gene Symbols FN1
Host Species Mouse
Immunogen Human fibronectin Purified from serum by affinity chromatography on gelatin-sepharose (Uniprot: P02751)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2335
Test Specificity Fibronectin is a dimeric glycoprotein of 440kDa, which is present in cells, extracellular matrix, and blood. It possesses at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectin is involved in cell adhesion, tissue organization, and wound healing. This monoclonal antibody is directed against the peptide core and reacts with both the plasma and cellular forms of fibronectin. It blocks the fibronectin-medicated cell attachment not by disrupting the collagen-fibronectin interaction, but by interfering with the attachment of fibronectin to its receptor on the cell surface.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.