Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibulin-3/EFEMP1 Antibody, Novus Biologicals™
SDP

Catalog No. NB300691005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300691005 0.05 mg
NB018172 0.025 mg
NB300691 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB300691005 Supplier Novus Biologicals Supplier No. NB3006910.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fibulin-3/EFEMP1 Polyclonal specifically detects Fibulin-3/EFEMP1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Fibulin-3/EFEMP1
Applications Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:100
Formulation Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide
Gene Alias DHRD, EGF containing fibulin-like extracellular matrix protein 1, EGF-containing fibulin-like extracellular matrix protein 1, Extracellular protein S1-5, FBLN3DRAD, FBNLFLJ35535, FIBL-3, fibrillin-like, Fibrillin-like protein, Fibulin-3, MGC111353, MLVT, MTLV, S1-5
Gene Symbols EFEMP1
Host Species Rabbit
Immunogen Synthetic cyclic peptide corresponding to N terminal amino acids 25-42 of Human EFEMP1, peptide was covalently modified. Sequence: TYTQCTDGYEWDPVRQQC
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 2202
Test Specificity The Anti EFEMP1 selective was generated against a conserved sequence near the N terminal region that is unique to EFEMP1 protein only. This detects a strong 62 kDa EFEMP1 protein in human.
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.