Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibulin-3/EFEMP1 Antibody (OTI1D9), HRP, Novus Biologicals™
SDP

Catalog No. NBP270700H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270700H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270700H Supplier Novus Biologicals Supplier No. NBP270700H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fibulin-3/EFEMP1 Monoclonal specifically detects Fibulin-3/EFEMP1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Fibulin-3/EFEMP1
Applications Western Blot
Classification Monoclonal
Clone OTI1D9
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias DHRD, EGF containing fibulin-like extracellular matrix protein 1, EGF-containing fibulin-like extracellular matrix protein 1, Extracellular protein S1-5, FBLN3DRAD, FBNLFLJ35535, FIBL-3, fibrillin-like, Fibrillin-like protein, Fibulin-3, MGC111353, MLVT, MTLV, S1-5
Gene Symbols EFEMP1
Host Species Mouse
Immunogen Full length recombinant protein of human Fibulin-3/EFEMP1 (NP_004096) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 2202.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.