Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ficolin-2 Antibody (16), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB355750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355750 Supplier Novus Biologicals Supplier No. NBP221613MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ficolin-2 Monoclonal antibody specifically detects Ficolin-2 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Ficolin-2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 16
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias 37 kDa elastin-binding protein, Collagen/fibrinogen domain-containing protein 2, EBP-37ficolin (collagen/fibrinogen domain-containing lectin) 2 (hucolin), FCNLficolin B, ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin), ficolin-2, Ficolin-B, Ficolin-beta, Hucolin, L-ficolin, P35, Serum lectin p35
Host Species Mouse
Immunogen Purified procaryot expressed recombinant human L-Ficolin and full-length L-Ficolin (Ficolin-2) protein.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2220
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.