Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ficolin-2 Antibody (19), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP105042UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105042UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105042UV Supplier Novus Biologicals Supplier No. NBP105042UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ficolin-2 Monoclonal specifically detects Ficolin-2 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Ficolin-2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 19
Conjugate DyLight 350
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 37 kDa elastin-binding protein, Collagen/fibrinogen domain-containing protein 2, EBP-37ficolin (collagen/fibrinogen domain-containing lectin) 2 (hucolin), FCNLficolin B, ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin), ficolin-2, Ficolin-B, Ficolin-beta, Hucolin, L-ficolin, P35, Serum lectin p35
Gene Symbols FCN2
Host Species Mouse
Immunogen Purified procaryot expressed recombinant human L-Ficolin and full-length L-Ficolin (Ficolin-2) protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2220
Test Specificity Reacts specifically with L-Ficolin. No cross reactivity is seen with M-Ficolin (Ficolin-1) and H-Ficolin (Ficolin-3).
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.