Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ficolin-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Ficolin-2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Ficolin-2 Polyclonal specifically detects Ficolin-2 in Human samples. It is validated for Western Blot.Specifications
Ficolin-2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
37 kDa elastin-binding protein, Collagen/fibrinogen domain-containing protein 2, EBP-37ficolin (collagen/fibrinogen domain-containing lectin) 2 (hucolin), FCN2, FCNL, FCNLficolin B, ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin), ficolin-2, Ficolin2, Ficolin-B, Ficolin-beta, Hucolin, L-Ficolin, P35, Serum lectin p35 | |
FCN2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q15485 | |
2220 | |
Mouse myeloma cell line NS0-derived recombinant human Ficolin-2 Leu26-Ala313 Accession # Q15485 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Ficolin-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant human Ficolin-3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title