Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ficolin-3 Antibody (4H5), Novus Biologicals™
SDP

Catalog No. NBP262788 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262788 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262788 Supplier Novus Biologicals Supplier No. NBP262788
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ficolin-3 Monoclonal specifically detects Ficolin-3 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Immunoassay.

Specifications

Antigen Ficolin-3
Applications Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay, Immunoassay
Classification Monoclonal
Clone 4H5
Concentration 0.1mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional, Immunoassay
Formulation PBS and 0.1% bovine serum albumin, 0.02 NaN3 with 0.02% Sodium Azide
Gene Alias Collagen/fibrinogen domain-containing lectin 3 p35, Collagen/fibrinogen domain-containing protein 3, FCNHficolin 3, ficolin (collagen/fibrinogen domain containing) 3 (Hakata antigen), ficolin (collagen/fibrinogen domain-containing) 3 (Hakata antigen), ficolin-3, HAKA1MGC22543, Hakata antigen, H-ficolin
Gene Symbols FCN3
Host Species Mouse
Immunogen Ficolin-3
Purification Method Protein G purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 8547
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.