Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ficolin-3 Antibody (FCN334), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP22373A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP22373A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP22373A30 Supplier Novus Biologicals Supplier No. NBP223573AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ficolin-3 Monoclonal specifically detects Ficolin-3 in Human samples. It is validated for ELISA.

Specifications

Antigen Ficolin-3
Applications ELISA
Classification Monoclonal
Clone FCN334
Conjugate Alexa Fluor 350
Dilution ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Collagen/fibrinogen domain-containing lectin 3 p35, Collagen/fibrinogen domain-containing protein 3, FCNHficolin 3, ficolin (collagen/fibrinogen domain containing) 3 (Hakata antigen), ficolin (collagen/fibrinogen domain-containing) 3 (Hakata antigen), ficolin-3, HAKA1MGC22543, Hakata antigen, H-ficolin
Gene Symbols FCN3
Host Species Mouse
Immunogen Recombinant human H-ficolin
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 8547
Test Specificity Clone FCN334 reacts specifically with H-ficolin. No cross-reaction has been observed with L- or M-ficolin
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.