Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIG4 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095247 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095247 20 μL
NB095246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095247 Supplier Novus Biologicals Supplier No. NBP30513020UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FIG4 Polyclonal specifically detects FIG4 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen FIG4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ALS11polyphosphoinositide phosphatase, CMT4J, dJ249I4.1, EC 3.1.3, EC 3.1.3.-, EC 3.1.3.36, FIG4 homolog (S. cerevisiae), FIG4 homolog, SAC1 lipid phosphatase domain containing (S. cerevisiae), hSac3, KIAA0274FIG4 homolog, SAC domain containing lipid phosphatase, Phosphatidylinositol 3,5-bisphosphate 5-phosphatase, Sac domain-containing inositol phosphatase 3, SAC domain-containing protein 3, SAC3FIG4 homolog, SAC1 domain containing lipid phosphatase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 608-907 of human FIG4 (NP_055660.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9896
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.