Learn More
Description
Specifications
Specifications
| Antigen | FIGLA |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 1:100 |
| Formulation | PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide |
| Gene Alias | BHLHC8, Class C basic helix-loop-helix protein 8, factor in the germline alpha, FIGalpha, folliculogenesis specific basic helix-loop-helix, Folliculogenesis-specific basic helix-loop-helix protein, POF6, Transcription factor FIGa |
| Gene Symbols | FIGLA |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
