Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIGNL1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB292376 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292376 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292376 Supplier Novus Biologicals Supplier No. NB1002356PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FIGNL1 Polyclonal antibody specifically detects FIGNL1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen FIGNL1
Applications Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias EC 3.6.4, EC 3.6.4.-, fidgetin-like 1, fidgetin-like protein 1
Host Species Rabbit
Immunogen Maps to a region between residue 325 and 375 of human Fidgetin-Like 1using the numbering given in entry NP_071399.2 (GeneID 63979).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63979
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.