Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIH-1/HIF-1AN Antibody (162c) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013811 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013811 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013811 Supplier Novus Biologicals Supplier No. NBP280738
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FIH-1/HIF-1AN Monoclonal specifically detects FIH-1/HIF-1AN in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen FIH-1/HIF-1AN
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 162c
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 5 μg/mL, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence : 1:25, Immunohistochemistry-Paraffin : 1:400
Formulation Tris-Glycine, 0.15 M NaCl
Gene Alias DKFZp762F1811, EC 1.14.11.16, factor inhibiting HIF1, Factor inhibiting HIF-1, FIH1FIH-1, FLJ20615, FLJ22027, hypoxia inducible factor 1, alpha subunit inhibitor, hypoxia-inducible factor 1-alpha inhibitor, Hypoxia-inducible factor asparagine hydroxylase, peptide-aspartate beta-dioxygenase
Gene Symbols HIF1AN
Host Species Mouse
Immunogen Full length human Factor Inhibiting HIF-1 protein [Swiss-Prot# Q9NWT6]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 55662
Test Specificity Human.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.